Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calred Solution, 0.02% w/v, Reagents
Click to view available options
Quantity:
1 L
Specifications
Specifications
Chemical Name or Material | Calred Solution, 0.02% w/v |
CAS | 7732-18-5, 3737-95-9 |
Boiling Point | 100°C |
Percent Purity | 0.02% |
Grade | Reagent |
Packaging | Glass Bottle |
Specific Gravity | 1.0002 |
Color | Purple |
Quantity | 1 L |
Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction