Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calreticulin Mouse anti-Human, Clone: CR213-2AG, Novus Biologicals™

Mouse Monoclonal Antibody

$213.45 - $352.89

Specifications

Antigen Calreticulin
Clone CR213-2AG
Concentration 1 mg/ml
Dilution Western Blot, ELISA
Applications Western Blot, ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB120551
View Documents
Novus Biologicals
NBP311672100UG
100 μg
Each for $352.89
Add to cart
 
NB120552
View Documents
Novus Biologicals
NBP31167250UG
50 μg
Each for $213.45
Add to cart
 
Description

Description

Calreticulin Monoclonal antibody specifically detects Calreticulin in Human samples. It is validated for Western Blot, ELISA
Specifications

Specifications

Calreticulin
1 mg/ml
Western Blot, ELISA
Purified
RUO
Human
autoantigen Ro, CALR, calregulin, calreticulin, cC1qR, CRP55, CRT, Endoplasmic reticulum resident protein 60, epididymis secretory sperm binding protein Li 99n, ERp60, FLJ26680, grp60, HACBP, HEL-S-99n, RO, Sicca syndrome antigen A (autoantigen Ro; calreticulin), SSA
Recombinant human calreticulin.
Monoclonal
Protein G purified
CR213-2AG
Western Blot, ELISA
Unconjugated
Mouse
Autophagy, Cancer, Cellular Markers, ER Markers, Immunology, Membrane Trafficking and Chaperones, Neurodegeneration, Neuroscience, Signal Transduction
0.2 um-filtered solution in PBS, pH 7.4
811
IgG Kappa
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit