Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calreticulin Mouse anti-Human, Mouse, Biotin, Clone: 1G6A7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP147518B

Catalog No. NBP147518B


Add to cart

Description

Description

Calreticulin Monoclonal antibody specifically detects Calreticulin in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Calreticulin
1G6A7
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Purified
CALR
A synthetic peptide corresponding to the C-terminus (EEEDVPGQAKDELC) of human Calreticulin, conjugated to KLH. [UniProt# P27797]
Ammonium sulfate precipitation
RUO
Store at 4C in the dark.
Monoclonal
Human, Mouse
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Biotin
PBS with 0.05% Sodium Azide
autoantigen Ro, calregulin, calreticulin, cC1qR, CRP55, CRT, CRTC, Endoplasmic reticulum resident protein 60, ERp60, FLJ26680, grp60, RO, Sicca syndrome antigen A (autoantigen Ro; calreticulin), SSAHACBP
Mouse
IgG2a
0.1 ml
Autophagy, Cancer, Cellular Markers, ER Markers, Membrane Trafficking and Chaperones, Neuroscience, Neurotransmission, Signal Transduction
Primary
811
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only