Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calreticulin Mouse anti-Human, Mouse, PE-Cyanine7, Clone: 1G6A7, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP147518PECY7
Description
Calreticulin Monoclonal antibody specifically detects Calreticulin in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
Calreticulin | |
1G6A7 | |
Flow Cytometry | |
Conjugated | |
CALR | |
A synthetic peptide corresponding to the C-terminus (EEEDVPGQAKDELC) of human Calreticulin, conjugated to KLH. [UniProt No. P27797] | |
Ammonium sulfate precipitation | |
Autophagy, Cancer, Cellular Markers, ER Markers, Membrane Trafficking and Chaperones, Neuroscience, Neurotransmission, Signal Transduction | |
Primary | |
811.0 |
Flow Cytometry | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
autoantigen Ro, calregulin, calreticulin, cC1qR, CRP55, CRT, CRTC, Endoplasmic reticulum resident protein 60, ERp60, FLJ26680, grp60, RO, Sicca syndrome antigen A (autoantigen Ro; calreticulin), SSAHACBP | |
Mouse | |
IgG2a | |
0.1 mL | |
Store at 4°C in the dark. Do not freeze. | |
Monoclonal | |
Human, Mouse |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title