Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calretinin Mouse anti-Human, Clone: CALB2/2602, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30736510UG
Description
Calretinin Monoclonal specifically detects Calretinin in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Protein Array.Specifications
Calretinin | |
Monoclonal | |
0.2 mg/ml | |
Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
29 kDa calbindin, CAB29, CAL2, calbindin 2, calbindin 2, (29kD, calretinin), calbindin 2, 29kDa (calretinin), calbindin D29K, calretinin, CR | |
Recombinant human Calretinin (Calbindin 2) protein fragment (around aa23-242) (exact sequence is proprietary) | |
100 μg | |
Cellular Markers, Phospho Specific | |
794 | |
Store at 4°C. | |
IgG1 κ |
Western Blot, Immunohistochemistry (Paraffin), Peptide Array | |
CALB2/2602 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title