Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calretinin Mouse anti-Human, Clone: CALB2/2602, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Calretinin
Clone CALB2/2602
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Applications Western Blot, Immunohistochemistry (Paraffin), Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122204
View Documents
Novus Biologicals
NBP30736520UG
20 μg
Each for $138.37
Add to cart
 
NB122203
View Documents
Novus Biologicals
NBP30736510UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

Calretinin Monoclonal specifically detects Calretinin in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Protein Array.
Specifications

Specifications

Calretinin
0.2 mg/ml
Western Blot, Immunohistochemistry (Paraffin), Peptide Array
Purified
RUO
Human
29 kDa calbindin, CAB29, CAL2, calbindin 2, calbindin 2, (29kD, calretinin), calbindin 2, 29kDa (calretinin), calbindin D29K, calretinin, CR
Recombinant human Calretinin (Calbindin 2) protein fragment (around aa23-242) (exact sequence is proprietary)
Monoclonal
Protein A purified
CALB2/2602
Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Unconjugated
Mouse
Cellular Markers, Phospho Specific
10 mM PBS with 0.05% BSA
794
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit