Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calretinin Mouse anti-Human, Monkey, Clone: 7H1G3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP261720

Catalog No. NBP2-61720


Add to cart

Description

Description

Calretinin Monoclonal antibody specifically detects Calretinin in Human, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry.
Specifications

Specifications

Calretinin
7H1G3
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000
Purified
CALB2
Purified recombinant fragment of human CALB2 (AA: 172-271) expressed in E. Coli.
31.5 kDa
0.1 ml
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Human, Monkey
ELISA, Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
PBS with 0.05% Sodium Azide
29 kDa calbindin, CAB29, CAL2, calbindin 2, calbindin 2, (29kD, calretinin), calbindin 2, 29kDa (calretinin), calbindin D29K, calretinin, CR
Mouse
IgG1
Protein G purified
Cellular Markers, Phospho Specific
Primary
794
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit