Learn More
Calretinin Rabbit anti-Human,Mouse,Rat, Unconjugated, Clone: 3K22, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma ZRB505425UL
 View more versions of this product
Catalog No. ZRB505425MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Calretinin Monoclonal antibody specifically detects Calretinin in Human,Mouse,Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications
Specifications
Calretinin
3K22
Unconjugated
CR
Rabbit
IgG
Protein A Purified
Neuroscience
Primary
Human, Mouse, Rat
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
120 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Purified
Calb2
GST/His-tagged recombinant fragment corresponding to 99 amino acids from the internal region of rat Calretinin.
∼30 kDa observed; 31.40 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
25 μL
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Monoclonal