Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calsyntenin-3 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00 - $501.50

Specifications

Antigen Calsyntenin-3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP15971320
SDP
View Documents
Novus Biologicals
NBP15971320UL
20 μL
Each for $206.00
Only null left
Add to Cart
 
NBP159713
SDP
View Documents
Novus Biologicals
NBP159713
100 μL
Each for $501.50
Only null left
Add to Cart
 
Description

Description

Calsyntenin-3 Polyclonal specifically detects Calsyntenin-3 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Calsyntenin-3
Polyclonal
Rabbit
Q9BQT9
9746
Synthetic peptides corresponding to CLSTN3 (calsyntenin 3) The peptide sequence was selected from the N terminal of CLSTN3)(50ug). Peptide sequence QICYYEILTPNTPFLIDNDGNIENTEKLQYSGERLYKFTVTAYDCGKKRA.
Primary
Western Blot
Unconjugated
RUO
alcadein beta, Alcadein-beta, Alc-beta, cadherin-related family member 14, calsyntenin 3, calsyntenin-3, CDHR14, CS3, CSTN3, KIAA0726alcbeta, MGC131797, MGC138488
CLSTN3
IgG
106 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.