Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CaM Kinase I Rabbit anti-Human, HRP, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP299907H

Catalog No. NB063445


Add to cart

Description

Description

CaM Kinase I Polyclonal specifically detects CaM Kinase I in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

CaM Kinase I
HRP
PBS
calcium/calmodulin-dependent protein kinase I, calcium/calmodulin-dependent protein kinase type 1, CaM kinase I, CaM kinase I alpha, caM-KI, CaMKI, CaMKI-alpha, EC 2.7.11, EC 2.7.11.17, MGC120317, MGC120318
Produced in rabbits immunized with purified, recombinant CaM Kinase I (NP_003647.1; Leu 2-Leu 370).
Affinity purified
RUO
Store at 4°C in the dark.
Polyclonal
Human
Western Blot, ELISA
Western Blot, ELISA
Purified
Rabbit
IgG
0.1 mL
Neuroscience, Protein Kinase, Sensory Systems, Vision
Primary
8536
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit