Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CaM Kinase II gamma Mouse anti-Human, FITC, Clone: 6A4A10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237269F

Catalog No. NBP237269F

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

CaM Kinase II gamma
6A4A10
ELISA
Purified
CAMK2G
Purified recombinant fragment of human CAMK2G (AA: 322-481) expressed in E. Coli.
Protein A or G purified
Protein Kinase, Signal Transduction, Tyrosine Kinases, Wnt Signaling Pathway
Primary
818
ELISA
FITC
PBS with 0.05% Sodium Azide
calcium/calmodulin-dependent protein kinase (CaM kinase) II gamma, calcium/calmodulin-dependent protein kinase II gamma, calcium/calmodulin-dependent protein kinase type II subunit gamma, CaM kinase II subunit gamma, CAMK, CAMKGFLJ16043, CAMK-II, CaMK-II subunit gamma, EC 2.7.11, EC 2.7.11.17, MGC26678
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit