Cambridge Diagnostic Products, Inc.™ Erado-Sol™ Stain Remover

Biodegradable, phosphate-free and pH neutral

Manufacturer: CAMBRIDGE DIAGNOSTIC PRODUCTS 20716

 View more versions of this product

Catalog No. 04-330-13

  Check Availability
Add to Cart

    Specifications

    Format Liquid
    Size (English) 16 oz.
    Size (Metric) 473.17mL
    Quantity Each
    Composition Ethanol 5 to 7%, Methanol <0.5%
    For Use With (Application) Removes most medical and histological stains from hands, glassware and lab equipment
    Product Type Erado-Sol™ Stain Remover

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit