Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ CAMK1 (CaMK1) Recombinant Human Protein
CAMK1 (CaMK1) is a serine⁄threonine kinase that is a member of the calcium⁄calmodulin-dependent protein kinase family.
Supplier: Thermo Scientific™ PV4427

Description
CAMK1 (CaMK1) is involved in cell differentiation, nervous system development, and signal transduction.Specifications
NP_003547 | |
1 mg | |
CAMK1 Kinase Family | |
His Tag | |
Ser⁄Thr Kinases: CAMK Group | |
Baculovirus | |
Full Length | |
Kinase Assay | |
Drug Development |
45.4 kDa | |
CAMK1 (CaMK1) | |
Yes | |
Storage Buffer: 50 mM Tris pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% Glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Frozen | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title