Learn More
Aviva Systems Biology Corp CAMK1D Antibody - middle region (ARP57405_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP57405_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CAMK1D. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CAMK1D. Alternate names: CKLiK, CaM-K1, CaMKIDApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62867