Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Camk2a, Mouse anti-Rat, R-PE, Clone: 6G9, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB18045

Catalog No. 89-337-970


Add to cart

Description

Description

Specifications

Specifications

Camk2a
6G9
ELISA Immunocytochemistry (1:50) Immunofluorescence (1:50) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:2000) Immunoprecipitation Radioimmunoassay Western Blot (1:10000) The optimal working dilution should be determined by the end user.
calcium/calmodulin-dependent protein kinase II alpha
PK2CDD|PKCCD
Mouse
IgG1
RUO
Antibody
Monoclonal
Rat
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Radioimmune Assays
R-PE
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide).
Liquid
Camk2a
Partially purified rat Camk2a.
100 μg
Store at -20°C.
Primary
25400
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit