Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Human CAMK2B (CaMKII Beta), His Tag Recombinant Protein
Human CAMK2B (CaMKII Beta), His Tag Recombinant Protein
$568.00 - $1914.00
Specifications
Accession Number | NP_742078 |
---|---|
For Use With (Application) | Kinase Assay |
Molecular Weight (g/mol) | 60.5 kDa |
Gene Alias | CAMK2B (CaMKII Beta), CAMK2 |
KinaseFamily | CAMK2 Kinase Family |
Description
As a dominant kinase in the central nervous system CAMK2B (CaMKII beta) may function in long term potentiation and neurotransmitter release.Specifications
NP_742078 | |
60.5 kDa | |
CAMK2 Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, and 50% Glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors |
Kinase Assay | |
CAMK2B (CaMKII Beta), CAMK2 | |
Human | |
His Tag | |
Ser⁄Thr Kinases: CAMK Group | |
Baculovirus |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title