Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAMK2G Mouse anti-Human, Rat, Clone: 8G10C1, Abnova™

Mouse monoclonal antibody raised against partial recombinant human CAMK2G.

Manufacturer:  Abnova Corporation MAB12257

Catalog No. 89-933-713


Add to cart

Description

Description

The product of this gene belongs to the Serine/Threonine protein kinase family, and to the Ca(2+)/calmodulin-dependent protein kinase subfamily. Calcium signaling is crucial for several aspects of plasticity at glutamatergic synapses. In mammalian cells the enzyme is composed of four different chains: alpha, beta, gamma, and delta. The product of this gene is a gamma chain. Six alternatively spliced variants that encode six different isoforms have been characterized to date. Additional alternative splice variants that encode different isoforms have been described, but their full-length nature has not been determined. [provided by RefSeq]

Specifications

Specifications

CAMK2G
8G10C1
Mouse monoclonal antibody raised against partial recombinant human CAMK2G.
In PBS (0.05% sodium azide)
CAMK/CAMK-II/CAMKG/FLJ16043/MGC26678
Mouse
IgG1
RUO
Primary
818
ELISA, Flow Cytometry, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
ELISA (1:10000) Flow Cytometry (1:200-1:400) Immunohistochemistry (1:200-1:1000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Liquid
CAMK2G
Recombinant protein corresponding to amino acids 322-481 of human CAMK2G.
100 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human, Rat
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit