Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CAMK1D Monoclonal Antibody (OTI2C6), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA525045
Description
CAMK1D Monoclonal antibody specifically detects CAMK1D in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
CAMK1D | |
Monoclonal | |
0.59 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8IU85 | |
CAMK1D | |
Full length human recombinant protein of CAMK1D produced in HEK293T cell | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
OTI2C6 | |
Unconjugated | |
CAMK1D | |
A630059D12Rik; calcium/calmodulin dependent protein kinase ID; calcium/calmodulin-dependent protein kinase 1D; calcium/calmodulin-dependent protein kinase ID; calcium/calmodulin-dependent protein kinase type 1D; CaM kinase I delta; caM kinase ID; CaM-K1; CAMK1D; CAMKI; caMKI delta; caM-KI delta; CAMKID; CaMKIdelta; CaMKI-delta; camKI-like protein kinase; CKLiK; E030025C11Rik; mCKLiK; MGC120317; MGC120318; OTTHUMP00000019119; OTTHUMP00000019120; OTTHUMP00000045030; RGD1560691; RP11-462F15.1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
57118 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction