Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CaMKIV Mouse anti-Human, DyLight 550, Clone: 8C5B8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237428R

Catalog No. NBP237428R

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

CaMKIV
8C5B8
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
CAMK4
Purified recombinant fragment of human CAMK4 (AA: 35-292 ) expressed in E. Coli.
Protein A or G purified
Cellular Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Protein Kinase
Primary
814
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
brain Ca(2+)-calmodulin-dependent protein kinase type IV, brain Ca++-calmodulin-dependent protein kinase type IV, calcium/calmodulin-dependent protein kinase IV, calcium/calmodulin-dependent protein kinase type IV, calcium/calmodulin-dependent protein kinase type IV catalytic chain, CAM kinase- GR, CAM kinase IV, CaM kinase-GR, caMK, CaMK IV, CaMK-GR, EC 2.7.11, EC 2.7.11.17, IV, MGC36771
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit