Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAMKK2 Mouse anti-Human, Mouse, Clone: 239CT7.5.3, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Thermo Scientific MA537515

Encompass

Catalog No. PIMA537515


Add to cart

Description

Description

CAMKK2 Monoclonal antibody specifically detects CAMKK2 in Human, Mouse samples. It is validated for Western Blot
Specifications

Specifications

CAMKK2
Monoclonal
Lot-Specific
PBS with 0.09% sodium azide
Q8C078, Q96RR4
6330570N16Rik; AW061083; Ca+/Calmodulin-dependent protein kinase kinase beta (CaM-kinase kinase beta); calcium/calmodulin dependent protein kinase kinase 2; calcium/calmodulin-dependent protein kinase 2 beta; calcium/calmodulin-dependent protein kinase beta; calcium/calmodulin-dependent protein kinase kinase 2; calcium/calmodulin-dependent protein kinase kinase 2, beta; calcium/calmodulin-dependent protein kinase kinase beta; caM-kinase kinase 2; CaM-kinase kinase beta; CAMKK; caMKK 2; caM-KK 2; caMKK beta; CaM-KK beta; CAMKK beta protein; Camkk2; CAMKKB; Kiaa0787; MGC15254; mKIAA0787
CAMKK2 recombinant protein
Euglobulin precipitation
RUO
Antibody
Monoclonal
Human, Mouse
Western Blot
239CT7.5.3
Unconjugated
CAMKK2
Liquid
Mouse
IgM, kappa
400 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
10645, 207565
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit