Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CAMKV Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101203
Description
Antibody detects endogenous levels of total CAMK5.
CaMKV does not appear to have detectable kinase activity.
Specifications
CAMKV | |
Polyclonal | |
Unconjugated | |
CAMKV | |
1G5; 1G5z; BB074618; BC017634; calcium/calmodulin dependent protein; CaM kinase like vesicle associated; CaM kinase-like vesicle-associated; CaM kinase-like vesicle-associated b; caM kinase-like vesicle-associated protein; CAMK5; Camkv; camkvb; DKFZp564G2082; hypothetical protein MGC63506; testis tissue sperm-binding protein Li 52e; VACAMKL; vesicle-associated calmodulin-binding protein; zgc:63506 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
235604, 79011, 79012 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q3UHL1, Q63092, Q8NCB2 | |
CAMKV | |
A synthesized peptide derived from human CAMKV(Accession Q8NCB2), corresponding to amino acid residues R384-A434. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction