Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CAMLG Mouse anti-Human, PE, Clone: OTI1A3, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP270331PE
Description
CAMLG Monoclonal antibody specifically detects CAMLG in Human samples. It is validated for Flow Cytometry.Specifications
CAMLG | |
OTI1A3 | |
Flow Cytometry | |
Affinity Purified, Conjugated | |
CAMLG | |
Full length recombinant protein of human CAMLG (NP_001736) produced in HEK293T cells. | |
Immunogen affinity purified | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Flow Cytometry | |
PE | |
PBS with 0.05% Sodium Azide | |
calcium modulating ligand, calcium signal-modulating cyclophilin ligand, calcium-signal modulating cyclophilin ligand, CAMLcalcium-modulating cyclophilin ligand, cyclophilin B-binding protein, MGC163197 | |
Mouse | |
IgG1 | |
0.1 mL | |
Primary | |
819.0 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title