Learn More
Cell Biolabs IncSupplier Diversity Partner CAMP ELISA KIT 5X96W COLORMETR  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner STA5005
cAMP ELISA Kit is a competitive enzyme immunoassay designed to measure cAMP in cell culture supernatants, plasma, serum, saliva, urine, and cell lysates. Read results in a standard plate reader. 5x96 wells including standards and unknowns.
Catalog No. 50-673-640