Learn More
Cell Biolabs IncSupplier Diversity Partner CAMP ELISA KIT 96W CHEMILUMIN  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner STA505
cGMP ELISA Kit is a competitive enzyme immunoassay designed to measure cGMP in cell culture supernatants, plasma, serum, saliva, urine, and cell lysates. Read results in a standard plate reader. 96 wells including standards and unknowns.
Catalog No. 50-673-642