Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Camphanic acid chloride, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Plastic bottle
39637-74-6
C10H13ClO3
216.66
MFCD00135626
PAXWODJTHKJQDZ-QVDQXJPCSA-N
1s-4,7,7-trimethyl-3-oxo-2-oxabicyclo 2.2.1 heptane-1-carbonyl chloride, --1s,4r-camphanoyl chloride, 1s---camphanic acid chloride
12364938
(4S)-1,7,7-trimethyl-2-oxo-3-oxabicyclo[2.2.1]heptane-4-carbonyl chloride
CC1(C2(CCC1(OC2=O)C(=O)Cl)C)C
39637-74-6
C10H13ClO3
216.66
MFCD00135626
PAXWODJTHKJQDZ-QVDQXJPCSA-N
1s-4,7,7-trimethyl-3-oxo-2-oxabicyclo 2.2.1 heptane-1-carbonyl chloride, --1s,4r-camphanoyl chloride, 1s---camphanic acid chloride
12364938
(4S)-1,7,7-trimethyl-2-oxo-3-oxabicyclo[2.2.1]heptane-4-carbonyl chloride
CC1(C2(CCC1(OC2=O)C(=O)Cl)C)C
Chiral derivatization reagent
Packaging Plastic bottle
Quantity 25g
Color Undesignated
Infrared Spectrum Authentic
Specific Rotation − 18.00
Melting Point 67.0°C to 71.0°C
Specific Rotation Condition − 18.00 (20.00°C c=2,CCl4)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Solubility Information Solubility in water: decomposes
Formula Weight 216.66
Physical Form Powder
Percent Purity 98%
Chemical Name or Material (-)-Camphanic acid chloride, 98%
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 254-552-4

TSCA : TSCA