Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (+/-)-Camphoric anhydride, 98%

Click to view available options
Quantity:
25g
50g
100g
595-30-2
C10H14O3
182.219
MFCD00067307
VFZDNKRDYPTSTP-UHFFFAOYSA-N
camphoric anhydride,d-camphoric anhydride,dl-camphoric anhydride,1,8,8-trimethyl-3-oxabicyclo 3.2.1 octane-2,4-dione,l-camphoric anhydride,--camphoric anhydride,+-1,2,2-trimethyl-1,3-cyclopentanedicarboxylic anhydride,1,3-cyclopentanedicarboxylic anhydride, 1,2,2-trimethyl-, +,3-oxabicyclo 3.2.1 octane-2,4-dione, 1,8,8-trimethyl-, 1s,5,8,8-trimethyl-3-oxabicyclo 3.2.1 octane-2,4-dione
94834
5,8,8-trimethyl-3-oxabicyclo[3.2.1]octane-2,4-dione
CC1(C2CCC1(C(=O)OC2=O)C)C
595-30-2
C10H14O3
182.219
MFCD00067307
VFZDNKRDYPTSTP-UHFFFAOYSA-N
camphoric anhydride,d-camphoric anhydride,dl-camphoric anhydride,1,8,8-trimethyl-3-oxabicyclo 3.2.1 octane-2,4-dione,l-camphoric anhydride,--camphoric anhydride,+-1,2,2-trimethyl-1,3-cyclopentanedicarboxylic anhydride,1,3-cyclopentanedicarboxylic anhydride, 1,2,2-trimethyl-, +,3-oxabicyclo 3.2.1 octane-2,4-dione, 1,8,8-trimethyl-, 1s,5,8,8-trimethyl-3-oxabicyclo 3.2.1 octane-2,4-dione
94834
5,8,8-trimethyl-3-oxabicyclo[3.2.1]octane-2,4-dione
CC1(C2CCC1(C(=O)OC2=O)C)C
Applications
(+/-)-Camphoric anhydride is useful in the synthesis of aliphatic-aromatic copolyesters. It is also used as pharmaceutical intermediates.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Store under dry inert gas. Store away from moisture and oxidizing agents. Protect from humidity and water.
Quantity 100g
Boiling Point 269°C to 271°C
Melting Point 220°C to 225°C
Beilstein 83114
Formula Weight 182.22
Percent Purity 98%
Chemical Name or Material (+/-)-Camphoric anhydride

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning