Learn More
Sklar InstrumentsSupplier Diversity Partner Campion Uterine Forceps, Sklar, 9-1/4 in., Curved, Serrated, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 907898
Campion Uterine Forceps, Sklar, 9-1/4 in., Curved, Serrated, Stainless Steel. Sklar Campion Uterine Forceps are a non-ratcheted, finger ring OB/GYN instrument. This grasping instrument features long shanks as well as long jaws, commonly seen in uterine forceps. It has a slight curvature, beginning at half the length of the jaws. The 15mm tips are a fenestrated, long, slightly cupped, oval shape. It has an overall length of 9-1/4 inches.
Catalog No. 50-143-6847