Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Campylobacter Fetus Mouse anti-Bacteria, Clone: 123/117, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10062424
Description
Campylobacter Fetus Monoclonal antibody specifically detects Campylobacter Fetus in Bacteria samples. It is validated for ELISA.Specifications
Campylobacter Fetus | |
Monoclonal | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
This product is specific for Campylobacter fetus (NCTC 10842, ATCC 27374), as confirmed in a simple ELISA. Campylobacter fetus is a gram negative bacteria that can cause systemic infections in immunocompromised hosts. It resists the normal bactericidal actions of serum due to its S-protein capsule which prevents C3b binding. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
ELISA | |
123/117 | |
ELISA 1:100-1:2000 | |
C. Fetus (NCTC 10842 ATCC 27374) | |
0.1 mg | |
Primary | |
Bacteria | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only