Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Campylobacter Jejuni Antibody (380/412), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10062426JF646
Description
Campylobacter Jejuni Monoclonal antibody specifically detects Campylobacter Jejuni in Bacteria samples. It is validated for ELISASpecifications
Campylobacter Jejuni | |
Monoclonal | |
Janelia Fluor 646 | |
Campilobacter | |
C. jejuni Penner 2 | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG2a |
ELISA | |
380/412 | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Bacteria | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction