Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Campylobacter Jejuni Mouse anti-Bacteria, Clone: 2E-10.- Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

$378.35

Specifications

Antigen Campylobacter Jejuni
Clone 2E-10.
Host Species Mouse
Isotype IgM Kappa
Gene Alias Campilobacter
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB254354100 View Documents Novus Biologicals
NBP254354100UG
100 ug Each for $378.35
Add to cart
 
NB254354200 View Documents Novus Biologicals
NBP254354200UG
200 ug This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Campylobacter Jejuni Monoclonal antibody specifically detects Campylobacter Jejuni in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Campylobacter Jejuni
Mouse
Campilobacter
ELISA, Immunocytochemistry, Immunofluorescence
Unconjugated
Store at -20 to -80C. Avoid freeze-thaw cycles.
Monoclonal
1 mg/mL
2E-10.
IgM Kappa
Bacteria
RUO
Supernatant
Primary
PBS with No Preservative

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit