Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Campylobacter Jejuni Mouse anti-Bacteria, Clone: 2E-10., Novus Biologicals™
Mouse Monoclonal Antibody
Specifications
Antigen | Campylobacter Jejuni |
---|---|
Clone | 2E-10. |
Concentration | 0.2 mg/mL |
Dilution | ELISA 1 - 5 ug/ml, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml |
Applications | ELISA, Immunocytochemistry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB253148200
|
Novus Biologicals
NBP253148200UG |
200 μg | Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. | N/A | |||||
Description
Campylobacter Jejuni Monoclonal antibody specifically detects Campylobacter Jejuni in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Campylobacter Jejuni | |
0.2 mg/mL | |
ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Bacteria | |
Campilobacter | |
IgM κ | |
Tissue culture supernatant |
2E-10. | |
ELISA 1 - 5 ug/ml, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml | |
Monoclonal | |
Supernatant | |
RUO | |
10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
Total sonicate of Campylobacter jejuni | |
Primary | |
Store at 4C. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title