Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CAMTA1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550638

Catalog No. PA550638


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total CAMTA1 protein.

CAMTA1 is a transcriptional activator. May act as a tumor suppressor.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CAMTA1
Polyclonal
Unconjugated
CAMTA1
1810059M14Rik; 2310058O09Rik; AI316882; AW987475; calmodulin binding transcription activator 1; calmodulin-binding transcription activator 1; Camta1; CANPMR; Kiaa0833; mKIAA0833; MSTP023
Rabbit
Antigen affinity chromatography
RUO
100072, 23261
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.9 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
A2A891, Q9Y6Y1
CAMTA1
Synthetic peptide corresponding to residues near the C terminal of human calmodulin binding transcription activator 1.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.