Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Canada balsam, natural filtered

Mounting medium for microscopy

Manufacturer:  Alfa Aesar™ A1628922

Quantity 100g

 View more versions of this product

Catalog No. AAA1628922


  Check Availability
Add to cart

Specifications

Description

Applications
Canada balsam, is used in film forming and masking. It is also used as a mounting media for microscopy. It is also used in chemical, Pharmaceutical research, pharmaceutical intermediate and research. Canada Balsam is used in the preparation of slide mounts. A turpentine made of resin which is dissolved in essential oils.

Solubility
It is soluble in benzene, chloroform, xylene, ethyl acetate, ether, oil of cedar, and oil of turpentine. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Air and moisture sensitive. Keep away from strong oxidizing agents.
Description & Specifications

Chemical Identifiers

8007-47-4
UN1993
Balsam Canada
MFCD00132800
14,946
Canada balsam, Natural filtered

Specifications

0.99g/mL
100g
Very Viscous Liquid
UN1993
Balsam Canada
1.5230
8007-47-4
MFCD00132800
14,946
Canada balsam, Natural filtered

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

RTECSNumber : CP2352500

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit