Canada balsam, natural filtered, Alfa Aesar™

Manufacturer: Alfa Aesar A1628936

Quantity 500g

 View more versions of this product

Catalog No. AAA1628936

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 8007-47-4
    MDL Number MFCD00132800
    UN Number UN1993
    Merck Index 14,946
    Synonym Balsam Canada
    Chemical Name or Material Canada balsam, natural filtered

    Specifications

    Quantity 500g
    MDL Number MFCD00132800
    CAS 8007-47-4
    Merck Index 14,946
    Health Hazard 2 GHS H Statement
    H226;
    Flammable liquid and vapour.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    TSCA TSCA
    UN Number UN1993
    Synonym Balsam Canada
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a;
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number CP2352500
    Flash Point 43°C (109°F)
    Chemical Name or Material Canada balsam, natural filtered

    Description

    Applications
    Canada balsam, is used in film forming and masking. It is also used as a mounting media for microscopy. It is also used in chemical, Pharmaceutical research, pharmaceutical intermediate and research. Canada Balsam is used in the preparation of slide mounts. A turpentine made of resin which is dissolved in essential oils.

    Solubility
    It is soluble in benzene, chloroform, xylene, ethyl acetate, ether, oil of cedar, and oil of turpentine. Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Air and moisture sensitive. Keep away from strong oxidizing agents.

    GHS H Statement
    H226;
    Flammable liquid and vapour.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a;
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit