Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Canadian Balsam, Neutral, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
5 lb.
50 lb.
Packaging:
Amber Glass Bottle
Fiber Drum
8007-47-4
8007-47-4
Quantity 500 g
CAS Max % 1
Packaging Amber Glass Bottle
Refractive Index 1.5190-1.5260
Specific Gravity 0.980 - 0.993
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.