Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cancer Diagnostics, Inc.™ Tissue Marking Dyes
Formulated to be used without a mordant
Supplier: Cancer Diagnostics, Inc.™ 07275
Description
- Dyes are pigment-based with an alcohol foundation for consistency and durability
- Proprietary blend of materials remains bright through processing and produces clean, crisp results
- Will not run or bleed on tissue
- Impervious to laboratory chemicals and reagents
Order Info
Individual colors available in two sizes; also available in systems with sets of six or seven colors, wood tray and 50 applicator sticks. Two-year shelf life.
Specifications
93°C | |
Yellow | |
Ea. | |
8.5 to 10 |
2 oz. | |
Tissue Marking Dye | |
Isopropyl Alcohol 400ppm, Ammonia 35ppm | |
Liquid |
Safety and Handling
ShelfLife : 2 Years
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction