Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cancer Diagnostics, Inc.™ Tissue Marking Dyes
Shop All Cancer Diagnostics Products

Click to view available options
Quantity:
2 oz.
8 oz.
Color:
Black
Blue
Green
Green, Black, Red, Yellow, Blue, Orange
Green, Black, Red, Yellow, Blue, Orange, Violet
Orange
Red
Yellow
Description
- Dyes are pigment-based with an alcohol foundation for consistency and durability
- Proprietary blend of materials remains bright through processing and produces clean, crisp results
- Will not run or bleed on tissue
- Impervious to laboratory chemicals and reagents
Order Info
Individual colors available in two sizes; also available in systems with sets of six or seven colors, wood tray and 50 applicator sticks. Two-year shelf life.
Specifications
Specifications
| Boiling Point | 93°C |
| Color | Yellow |
| Product Type | Tissue Marking Dye |
| Quantity | 8 oz. |
| Formulation | Isopropyl Alcohol 400ppm, Ammonia 35ppm |
| pH | 8.5 to 10 |
| Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction