Learn More
Cancer Diagnostics, Inc. Vortex Biopsy Micro Cassettes
Used for routine processing and embedding of tissues and cells during histological studies
Manufacturer: Cancer Diagnostics, Inc. VMT1004
Description
- Rounded micro hole interior basket without corners
- Filtration enhanced by side vents
- Two-piece lid design
- Processed biopsies are easy to find
- Various colors available
- Tubed, loose and taped options for packaging
Specifications
Taped | |
Pink | |
Biopsy Micro Cassette |