Learn More
Cancer Diagnostics, Inc.™ Vortex™ Biopsy Micro Cassettes
Used for routine processing and embedding of tissues and cells during histological studies
Manufacturer:  Cancer Diagnostics, Inc.™ VMSL1005
 View more versions of this product
Catalog No. 22-500-884
Add to cart
Description
Description
  • Rounded micro hole interior basket without corners
  • Filtration enhanced by side vents
  • Two-piece lid design
  • Processed biopsies are easy to find
  • Various colors available
  • Tubed, loose and taped options for packaging
Specifications
Specifications
Tubed
Yellow
Biopsy Micro Cassette