Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Candesartan cilexetil, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
145040-37-5
C33H34N6O6
610.671
GHOSNRCGJFBJIB-UHFFFAOYSA-N
candesartan cilexetil,atacand,amias,parapres,kenzen,candesartan cilexetil usan,candesartan cilexitil,candesartan cilextil,chembl1014
2540
CHEBI:3348
1-cyclohexyloxycarbonyloxyethyl 2-ethoxy-3-[[4-[2-(2H-tetrazol-5-yl)phenyl]phenyl]methyl]benzimidazole-4-carboxylate
CCOC1=NC2=CC=CC(=C2N1CC3=CC=C(C=C3)C4=CC=CC=C4C5=NNN=N5)C(=O)OC(C)OC(=O)OC6CCCCC6
This item is not returnable. View return policy
145040-37-5
C33H34N6O6
610.671
GHOSNRCGJFBJIB-UHFFFAOYSA-N
candesartan cilexetil,atacand,amias,parapres,kenzen,candesartan cilexetil usan,candesartan cilexitil,candesartan cilextil,chembl1014
2540
CHEBI:3348
1-cyclohexyloxycarbonyloxyethyl 2-ethoxy-3-[[4-[2-(2H-tetrazol-5-yl)phenyl]phenyl]methyl]benzimidazole-4-carboxylate
CCOC1=NC2=CC=CC(=C2N1CC3=CC=C(C=C3)C4=CC=CC=C4C5=NNN=N5)C(=O)OC(C)OC(=O)OC6CCCCC6

Prodrug of candesartan (Cat. No. 4791), an AT1 receptor antagonist

Prodrug of the angiotensin II receptor 1 (AT1) antagonist candesartan (Cat. No. 4791), which exhibits antihypertensive effects in animal models.
Quantity 50mg
Formula Weight 610.66
Percent Purity >99%
Chemical Name or Material Candesartan cilexetil

Recommended Storage : Store at +4°C