Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Canine CXCL8/IL-8 ELISA Kit (Colorimetric)

High sensitivity and specificity for detection of dog IL-8.

$602.00

Specifications

Product Type ELISA Kit (Colorimetric)
Assay Range 0.312 to 20ng/mL
Species Canine
Sensitivity 0.078ng/mL
Sample Type Serum, plasma, cell culture supernates, tissue homogenates
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB599671KT View Documents Novus Biologicals™
NBP2599671KIT
1 Kit Each for $602.00
Add to cart
 
Description

Description

The Canine CXCL8/IL-8 ELISA Kit (Colorimetric) measures canine CXCL8/IL-8 in serum, plasma, cell culture supernates, tissue homogenates.
Specifications

Specifications

ELISA Kit (Colorimetric)
Canine
Serum, plasma, cell culture supernates, tissue homogenates
RUO
For detection of dog IL-8
Store the unopened product at 2 to 8°C Protect from light. Do not use past expiration date.
Assay plate (12 x 8 coated Microwells), Biotin-antibody Diluent, Biotin-antibody (100 x concentrate), Adhesive Strip (For 96 wells), HRP-avidin (100 x concentrate), HRP-avidin Diluent ,Wash Buffer (25 x concentrate), TMB Substrate, Standard (Freeze dried), Sample Diluent, Stop Solution, Instruction manual
0.312 to 20ng/mL
0.078ng/mL
Immunology
50 to 100μL
3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8SCYB8, Emoctakin, GCP-1TSG-1, interleukin 8, K60, LECT, MDNCFb-ENAP, member 8, MONAPGCP1, NAP-1NAP1, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor
CXCL8/IL-8
ELISA

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit