Learn More
Canine distemper virus Mouse anti-Virus, Clone: HM195, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB13539
Catalog No. 89-033-502
Add to cart
Description
Description
Mouse monoclonal antibody raised against canine distemper virus.
  • High specificity
  • High reactivity
Specifications
Specifications
Canine distemper virus
HM195
Mouse monoclonal antibody raised against canine distemper virus.
In PBS, pH 7.4 (0.09% sodium azide).
Mouse
Protein A purification
RUO
Primary
This antibody reacts with surface envelope antigen of canine distemper virus.
ELISA, Lateral Flow
Unconjugated
ELISA Lateral-Flow Immunochromatographic Assay The optimal working dilution should be determined by the end user.
Liquid
IgG2a
1 mg
Store at 4°C.
Monoclonal
Virus
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)