Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Canine Folic acid ELISA Kit (Colorimetric)

High sensitivity and specificity for detection of canine folic acid.

$732.50

Specifications

Product Type ELISA Kit (Colorimetric)
Assay Range 0.195 to 50ng/mL
Species Canine
Sensitivity 0.195ng/mL
Sample Type Serum, plasma
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB599971KT View Documents Novus Biologicals™
NBP2599971KIT
1 Kit Each for $732.50
Add to cart
 
Description

Description

The Canine Folic acid ELISA Kit (Colorimetric) measures canine Folic acid in serum, plasma.
Specifications

Specifications

ELISA Kit (Colorimetric)
Canine
Serum, plasma
RUO
For detection of canine folic acid
Store the unopened product at 2 to 8°C Protect from light. Do not use past expiration date.
Instruction manual, Sample Diluen, Stop Solution, Standard (Freeze dried), Wash Buffer (25 x concentrate), TMB Substrate, HRP-conjugate (100 x concentrate), HRP-conjugate Diluent, Adhesive Strip (For 96 wells), Antibody Diluent, Antibody (100 x concentrate), Assay plate (12 x 8 coated Microwells)
0.195 to 50ng/mL
0.195ng/mL
Lipid and Metabolism
50 to 100μL
5 Methyl THFA, 5 Methyltetrahydrofolate, Folate, Tetrahydrofolate, Tetrahydrofolic acid, THFA
Folic acid
ELISA

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit