Learn More
Raybiotech IncSupplier Diversity Partner Canine IFN alpha ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELCIFNA1
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.18 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Canine IFN alpha.
Catalog No. 50-194-8885