Learn More
Raybiotech IncSupplier Diversity Partner Canine IL-8 (CXCL8) ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELCIL81
For cell culture supernatants, plasma, and serum samples. Sensitivity: 6 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detect canine IL-8. Other species not determined yet.
Catalog No. 50-194-8893