Learn More
Canine parvovirus Mouse anti-Virus, Clone: HM194, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB13635
Catalog No. 89-033-598
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Mouse monoclonal antibody raised against canine parvovirus.
  • High specificity
  • High reactivity
Specifications
Specifications
Canine parvovirus
HM194
Mouse monoclonal antibody raised against canine parvovirus.
In PBS, pH 7.4 (0.09% sodium azide).
Mouse
Protein A purification
RUO
Primary
Virus
ELISA, Lateral Flow
Unconjugated
ELISA Lateral-Flow Immunochromatographic Assay The optimal working dilution should be determined by the end user.
Liquid
IgG2a
1 mg
Store at 4°C.
Monoclonal
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)