Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Canine Progesterone ELISA Kit (Colorimetric)
High sensitivity and specificity for detection of canine progesterone.
Supplier: Novus Biologicals™ NBP2601231KIT
Description
The Canine Progesterone ELISA Kit (Colorimetric) measures canine Progesterone in serum, plasma, tissue homogenates.Specifications
0.125 to 50ng/mL | |
ELISA Kit (Colorimetric) | |
For detection of canine PROG | |
ELISA | |
1 Kit | |
Signal Transduction | |
Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date. | |
Progesterone |
0.125ng/mL | |
Plasma, Serum, Tissue Homogenates | |
Canine | |
Adhesive Strip (For 96 wells), Antibody, Assay plate, HRP-conjugate, Substrate A, Substrate B, Wash Buffer (20 x concentrate), Stop Solution, Standard, Instruction manual | |
RUO | |
50 to 100 μL | |
Pregn 4 ene 3 20 dione |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?
Provide Content Correction