Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Canine Thyroxine ELISA Kit (Colorimetric)
High sensitivity and specificity for detection of canine T4.
Supplier: Novus Biologicals™ NBP2601601KIT
Description
The Canine Thyroxine ELISA Kit (Colorimetric) measures canine Thyroxine in serum, plasma, cell culture supernates, tissue homogenates.Specifications
20 to 320ng/mL | |
ELISA Kit (Colorimetric) | |
For detection of canine T4 | |
ELISA | |
1 Kit | |
Signal Transduction | |
Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date. | |
Thyroxine |
20ng/mL | |
Cell Culture Supernatant, Plasma, Serum, Tissue Homogenates | |
Canine | |
Instruction manual, StandardStop, SolutionWash, Buffer (20 x concentrate), Substrate A, Substrate B, HRP-avidin, Conjugate, Assay plate, Adhesive Strip (For 96 wells) | |
RUO | |
50 to 100 μL | |
T4 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?
Provide Content Correction