Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Canine Thyroxine ELISA Kit (Colorimetric)

High sensitivity and specificity for detection of canine T4.

$501.50

Specifications

Product Type ELISA Kit (Colorimetric)
Assay Range 20 to 320ng/mL
Species Canine
Sensitivity 20ng/mL
Sample Type serum, plasma, cell culture supernates, tissue homogenates
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB601601KT View Documents Novus Biologicals™
NBP2601601KIT
1 Kit Each for $501.50
Add to cart
 
Description

Description

The Canine Thyroxine ELISA Kit (Colorimetric) measures canine Thyroxine in serum, plasma, cell culture supernates, tissue homogenates.
Specifications

Specifications

ELISA Kit (Colorimetric)
Canine
serum, plasma, cell culture supernates, tissue homogenates
RUO
For detection of canine T4
Store the unopened product at 2 to 8°C Protect from light. Do not use past expiration date.
Instruction manual, StandardStop, SolutionWash, Buffer (20 x concentrate), Substrate A, Substrate B, HRP-avidin, Conjugate, Assay plate, Adhesive Strip (For 96 wells)
20 to 320ng/mL
20ng/mL
Signal Transduction
50 to 100μL
T4
Thyroxine
ELISA

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit