Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Canine Toxoplasmosis (TOXO) Antibody (IgG) ELISA Kit (Colorimetric)

High sensitivity and specificity for detection of dog anti-toxoplasmosis antibody (IgG).

$732.50

Specifications

Product Type ELISA Kit (Colorimetric)
Assay Range Qualitative
Species Canine
Sample Type Serum
Research Discipline Cell Biology
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB601711KT View Documents Novus Biologicals™
NBP2601711KIT
1 Kit Each for $732.50
Add to cart
 
Description

Description

The Canine Toxoplasmosis (TOXO) Antibody (IgG) ELISA Kit (Colorimetric) measures canine Toxoplasmosis (TOXO) Antibody (IgG) in serum.
Specifications

Specifications

ELISA Kit (Colorimetric)
Canine
Cell Biology
50 to 100μL
Store the unopened product at 2 to 8°C Protect from light. Do not use past expiration date.
Adhesive Strip (For 96 wells), HRP-conjugate, Coated assay plate, Wash Buffer (20 x concentrate), Substrate A, Substrate B, Stop Solution, Instruction manual, Positive Control, Negative Control
Qualitative
Serum
RUO
For detection of dog anti-toxoplasmosis antibody (IgG)
Toxoplasmosis (TOXO) Antibody (IgG)
ELISA

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit